Preparative sorbents, LiChroprep®
LiChroprep®Si 60, 25 - 40 µm, 15, 1 kg
Supplier: MERCK PRODUCTION CHEMICALS
Preparative sorbents, LiChroprep®
Catalog # 1.09390.1000
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Jump to:
Product Details & Documents
LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.
- Reproducible homogeneous silica gel matrix
- Narrow, well defined particle size distribution for high performance and high permeability
- Excellent selectivity and efficiency
- Large number of applications
- Large batch size
- Comprehensive regulatory documents are available
LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.
Specifications
Specifications
- Particle Size:25 - 40 µm
- Pk:1 kg
- Pore size:15
- Description:LiChroprep®Si 60



