Order Entry
Italy
Orders LinkContactUsLinkComponent
Preparative sorbents, LiChroprep®

Preparative sorbents, LiChroprep®

LiChroprep® Si 60, 15 - 25 µm, 14, 1 kg
Catalog # 1.09336.1000
Preparative sorbents, LiChroprep®
Catalog # 1.09336.1000
CAS Number:  
Preparative sorbents, LiChroprep®

LiChroprep® Si 60, 15 - 25 µm, 14, 1 kg

CAS Number:  
Price
671,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.

  • Reproducible homogeneous silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Excellent selectivity and efficiency
  • Large number of applications
  • Large batch size
  • Comprehensive regulatory documents are available

LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.

Specifications

Specifications

  • Particle Size:
    15 - 25 µm
  • Pk:
    1 kg
  • Pore size:
    14
  • Description:
    LiChroprep® Si 60

Specifications