Preparative sorbents, LiChroprep®
LiChroprep® RP-18, 25 - 40 µm, 04, 100 g
Supplier: MERCK PRODUCTION CHEMICALS
Preparative sorbents, LiChroprep®
Catalog # 1.09303.0100
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Jump to:
Product Details & Documents
LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.
- Reproducible homogeneous silica gel matrix
- Narrow, well defined particle size distribution for high performance and high permeability
- Excellent selectivity and efficiency
- Large number of applications
- Large batch size
- Comprehensive regulatory documents are available
LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.
Documents
Specifications
Specifications
- Particle Size:25 - 40 µm
- Pk:100 g
- Pore size:04
- Description:LiChroprep® RP-18
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



