Order Entry
Italy
ContactUsLinkComponent
SGE SYRINGE 500µL REMOVABLE NEEDLE IS 25 GAUGE (0.50 MM OD) 50 MM LONG WITH A BEVEL POINT STYLE. BARREL HAS A PTFE SEALING GLAND AT THE REAR FLANGE
Catalog # SGEA007610
SGE SYRINGE 500µL REMOVABLE NEEDLE IS 25 GAUGE (0.50 MM OD) 50 MM LONG WITH A BEVEL POINT STYLE. BARREL HAS A PTFE SEALING GLAND AT THE REAR FLANGE
Catalog # SGEA007610
Frequently Bought Together