Order Entry
Italy
ContactUsLinkComponent
SGE SYRINGE 50µL REMOVABLE NEEDLE IS STAINLESS STEEL 0.50 MM OD (25 GAUGE) 50 MM LONG AND HAS A BEVEL POINT STYLE. REAR FLANGE IS TH READED AND HAS A PTFE SEAL FOR A GAS TIGHT SEAL
Codice catalogo SGEA004610
SGE SYRINGE 50µL REMOVABLE NEEDLE IS STAINLESS STEEL 0.50 MM OD (25 GAUGE) 50 MM LONG AND HAS A BEVEL POINT STYLE. REAR FLANGE IS TH READED AND HAS A PTFE SEAL FOR A GAS TIGHT SEAL
Codice catalogo SGEA004610
Frequently Bought Together