Order Entry
Italy
ContactUsLinkComponent
SGE SYRINGE 25µL FIXED NEEDLE CTC BARREL 6.5MM NEEDLE IS STAINLESS STEEL 0.72 MM OD (22 GAUGE) 51 MM LONG WITH LC POINT STYLE
Catalog # SGEA003984
SGE SYRINGE 25µL FIXED NEEDLE CTC BARREL 6.5MM NEEDLE IS STAINLESS STEEL 0.72 MM OD (22 GAUGE) 51 MM LONG WITH LC POINT STYLE
Catalog # SGEA003984
Frequently Bought Together