Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Microvolume syringes 5 µl

26, 5 µl, 50 mm, 0,47 mm, Bevelled, 5R-GP
Catalog # 549-0273
Microvolume syringes 5 µl
Catalog # 549-0273
Microvolume syringes 5 µl

26, 5 µl, 50 mm, 0,47 mm, Bevelled, 5R-GP

Price
164,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

These are plunger-in-barrel syringes. Each plunger is individually fitted for each microsyringe for perfect sealing.

  • Longer life
  • Superior performance and robustness
  • Reduced carryover

Plunger protection incorporates a plunger protection guide and plunger button reinforcement to minimise the risk of bent plungers.

Guided plunger microvolume syringes have an extended barrel and reinforced plunger that provides additional protection.

* SuperfleX, with flexible plunger. SuperfleX microvolume syringes use a super elastic alloy plunger that will not bend or kink.

Specifications
  • Ext.Ø:
    0,47 mm
  • Gauge:
    26
  • Type:
    5R-GP
  • Type of tip:
    Bevelled
  • Volume:
    5 µl
  • Length:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.