Jump to:
Product Details & Documents
- CAS Number:1310-65-2
- Formula:LiOH
- MDL Number:MFCD00011095
- Molecular Weight:23.95 g/mol
- Merck Index:13,05556
- Boiling Pt:924 °C (1013 hPa)
- Melting Pt:462 °C
- Density:1.46 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-183-4
- ADR:8,II
- :M-Clarity™ quality level = MQ300
- UN:2680
Documents
msds_105691
SDS
ProductPage_105691
Product specification
Specifications
Specification Test Results
- Solubility:71 g/l (20 °C)
- Melting Point:462 °C
- Molar Mass:23.95 g/mol
- Bulk Density:550 kg/m3
- Density:2.54 g/cm3
- pH:12 (50 g/l, H2O, 50 °C)
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



