Cyclohexanone, EMPLURA®, extra pure, Supelco®
Stainless steel drum, returnable, 10 L
: Merck
Cyclohexanone, EMPLURA®, extra pure, Supelco®
1.02888.6010
: Merck
: 108-94-1
: Pimelic Ketone
- :108-94-1
- :C₆H₁₀(=O)
- :MFCD00001625
- :98.14 g/mol
- :13,02755
- :155.6 °C (1013 hPa)
- :–31 °C
- :0.9478 g/cm³ (20 °C)
- :43 °C
- :1915
- :3,III
- :M-Clarity™ quality level = MQ200
- :Ambient
- :203-631-1
- :01-2119453616-35
- Pack type:Stainless steel drum, returnable
- Pk:10 L
- Cat. No.:1.02888.6010
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



