Order Entry
Italy
Orders LinkContactUsLinkComponent
Seals for threaded connection caps (Septa)

Seals for threaded connection caps (Septa)

GL25 closure size, 22 mm, 2 mm
Catalog # 201-1963
Supplier:   BIRZLE PETER
Seals for threaded connection caps (Septa)
Catalog # 201-1963
Supplier:  BIRZLE PETER
Seals for threaded connection caps (Septa)
Seals for threaded connection caps (Septa)

GL25 closure size, 22 mm, 2 mm

Price
2,10 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Silicone, transparent.

  • Ideal for use as septa (for puncturing)
  • Very good chemical resistance
  • Very good thermal resistance

For threaded connection caps with GL thread.

Specifications
  • Ø:
    22 mm
  • For:
    GL25 closure size
  • Seq:
    03
  • Thickness:
    2 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.