Order Entry
Italy
Orders LinkContactUsLinkComponent
Seals for threaded connection caps (Septa)

Seals for threaded connection caps (Septa)

GL18 closure size, 16 mm, 2 mm
Catalog # 201-1962
Supplier:   BIRZLE PETER
Seals for threaded connection caps (Septa)
Catalog # 201-1962
Supplier:  BIRZLE PETER
Seals for threaded connection caps (Septa)
Seals for threaded connection caps (Septa)

GL18 closure size, 16 mm, 2 mm

Price
2,90 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Silicone, transparent.

  • Ideal for use as septa (for puncturing)
  • Very good chemical resistance
  • Very good thermal resistance

For threaded connection caps with GL thread.

Specifications
  • Ø:
    16 mm
  • For:
    GL18 closure size
  • Seq:
    02
  • Thickness:
    2 mm