Buckets, Nalgene®
White, 13,2 L
Catalog # 216-0095
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, white.
- Graduated
- Very good thermal and chemical resistance
- Autoclavable
Universal multipurpose bucket without spout. Nylon coated handle provides comfortable sturdy grip.
Specifications
- Capacity:13,2 L
- Colour:White
- Cat. No.:216-0095
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact