Order Entry
Italy
Orders LinkContactUsLinkComponent
Buckets, Nalgene®
Buckets, Nalgene®
White, 7,6 L
Catalog # 216-2127
   Low Manufacturing Impact
Buckets, Nalgene®
Catalog # 216-2127
   Low Manufacturing Impact
Buckets, Nalgene®
Buckets, Nalgene®
White, 7,6 L
Price
81,60 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
PP, white.
  • Graduated
  • Very good thermal and chemical resistance
  • Autoclavable
Universal multipurpose bucket without spout. Nylon coated handle provides comfortable sturdy grip.
Specifications
  • Capacity:
    7,6 L
  • Colour:
    White
  • Cat. No.:
    216-2127
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact