Order Entry
Italy
ContactUsLinkComponent
Conductivity test solution LF 995 in FIOLAX® ampoules, 3 x 6 ampoules, test solution potassium chloride 0.01/0.1/1 mol/l (1.41/12.9/ 112 mS/cm) 1 * 18 Ampoul
Catalog # 663-4007
Supplier:  SI Analytics
Conductivity test solution LF 995 in FIOLAX® ampoules, 3 x 6 ampoules, test solution potassium chloride 0.01/0.1/1 mol/l (1.41/12.9/ 112 mS/cm) 1 * 18 Ampoul
Catalog # 663-4007
Supplier:  SI Analytics
Frequently Bought Together