Order Entry
Italy
Orders LinkContactUsLinkComponent
Closed dome tanks
Closed dome tanks
Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm, 380 l, 724×1321 mm
Catalog # NALG2650-0100
   Low Manufacturing Impact
Closed dome tanks
Catalog # NALG2650-0100
   Low Manufacturing Impact
Closed dome tanks
Closed dome tanks
Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm, 380 l, 724×1321 mm
Price
3 450,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
These dome tanks are designed for use as closed containment systems. Tanks feature a 150 mm neck opening fitted with a gasketed closure for secure sealing.
Documents
Specifications
  • Capacity:
    380 l
  • Description:
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
  • Ø ext.×H:
    724×1321 mm
  • Cat. No.:
    NALG2650-0100
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Cap Size
    For bottles
    Availability
    Price
  • with silicone gasket, true union clamp
    with silicone gasket, true union clamp
    NALG2651-0200
    152,3 mm
    75/115/ 210/380 l
    Log in for availability
    1 * 1 pezzi - 1 220,00 €