Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
Sample vials, clear, with screw thread, KIMBLE®

Sample vials, clear, with screw thread, KIMBLE®

30,00 ml, 27×75 mm, trasparente, Yes
Codice catalogo SCERC40752700D0C2
Nome del fornitore:   DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Codice catalogo SCERC40752700D0C2
Nome del fornitore:  DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Sample vials, clear, with screw thread, KIMBLE®

30,00 ml, 27×75 mm, trasparente, Yes

Price
124,00 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.

Specifications
  • Tappo incluso:
    Yes
  • Ø×A:
    27×75 mm
  • Capacità:
    30,00 ml
  • Colore:
    trasparente
  • Seq:
    06