Sample vials, clear, with screw thread, KIMBLE®
15,00 ml, 27×45 mm, trasparente, Yes
SCERC40452700B5C2
Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.
Documents
- Tappo incluso:Yes
- Ø×A:27×45 mm
- Capacità:15,00 ml
- Colore:trasparente
- Seq:04



