Sample vials, clear, with screw thread, KIMBLE®
5,00 ml, 20×41 mm, Clear, Yes
Codice catalogo SCERC40412000A5B2
Nome del fornitore: DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Codice catalogo SCERC40412000A5B2
Nome del fornitore: DWK Life Sciences
Jump to:
Product Details & Documents
Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.
Documents
Specifications
- Cap included:Yes
- Ø×H:20×41 mm
- Capacity:5,00 ml
- Colour:Clear
- Seq:02



