Order Entry
Italy
Orders LinkContactUsLinkComponent
Caps, LABOCAP, with grip
Caps, LABOCAP, with grip
LABOCAP, blue, 17-18 mm
Catalog # 391-5931
Supplier:   LUDI
Caps, LABOCAP, with grip
Catalog # 391-5931
Supplier:  LUDI
Caps, LABOCAP, with grip
Caps, LABOCAP, with grip
LABOCAP, blue, 17-18 mm
Price
24,30 €
Log in for availability
1 * 10 pezzi
Product Details & Documents
Aluminium.
  • Made of anodised aluminium with inner stainless steel spring
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
With grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
Specifications
  • For Ø ext.:
    17-18 mm
  • Description:
    LABOCAP, blue