Caps, LABOCAP, with grip
LABOCAP, silver, 17-18 mm
Codice catalogo 391-5930
Nome del fornitore: LUDI
Jump to:
Product Details & Documents
Aluminium.
- Made of anodised aluminium with inner stainless steel spring
- Springs clamp against the outer wall of the vessel
- Anodised and coloured
- Autoclavable
With grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
Specifications
- For Ø ext.:17-18 mm
- Description:LABOCAP, silver