Order Entry
Italy
Orders LinkContactUsLinkComponent
Inoculating loops, PS, Looplast®
Inoculating loops, PS, Looplast®
Inoculating loop, Looplast®, PS, shockproof, sterile, 10 µl, Blue, Bags of 20
Catalog # LPIT131018
Supplier:   LP ITALIANA
Inoculating loops, PS, Looplast®
Catalog # LPIT131018
Supplier:  LP ITALIANA
Inoculating loops, PS, Looplast®
Inoculating loops, PS, Looplast®
Inoculating loop, Looplast®, PS, shockproof, sterile, 10 µl, Blue, Bags of 20
Price
286,00 €
Log in for availability
1 * 2.000 pezzi
Specifications
  • Packed:
    Bags of 20
  • Capacity:
    10 µl
  • Colour:
    Blue
  • Description:
    Inoculating loop, Looplast®, PS, shockproof, sterile