Order Entry
Italy
Orders LinkContactUsLinkComponent
Weighing bottles, PP

Weighing bottles, PP

190 ml, 59×88 mm
Catalog # 611-9405
Supplier:   KARTELL
Weighing bottles, PP
Catalog # 611-9405
Supplier:  KARTELL
Weighing bottles, PP
Weighing bottles, PP

190 ml, 59×88 mm

Price
7,50 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

PP with lid.

  • Very good chemical resistance
  • Very good temperature resistance

Cylindrical shape, with knobbed lid.

Specifications
  • Ø×H:
    59×88 mm
  • Capacity:
    190 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.