Double bossheads
Double bosshead, thumb screw M8, Aluminium, 27 mm
BOCH5352
: BOCHEM
Double bossheads, either made of stainless steel or aluminium.
- Angle: 180 + 90°
- Different screw types available
- Description:Double bosshead, thumb screw M8
- Material:Aluminium
- For rod Ø:27 mm



