Order Entry
Italy
Orders LinkContactUsLinkComponent
Drug test kit
Catalog # 321652Q
Drug test kit
Catalog # 321652Q
Supplier:  
Drug test kit
Drug test kit
Discontinued
Product Details & Documents
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
  • Kit comprises:
  • 20 (4 boxes of 5, 0,2 ml) ampoules of Marquis reagent
  • 1×10 ml cocaine reagent
  • 1×10 ml barbiturate reagent
  • 1×10 ml LSD reagent
  • 1×20 ml cannabis reagent 1
  • 1×10 ml cannabis reagent 2
  • 1×5 g cannabis special solid reagent
  • 1×20 g neutraliser
  • 4 boxes of Whatman filter papers
  • 1 microspatula
  • 20 ampoule breakers
  • 25 disposable spatulas
  • forceps
  • 4 watch glasses
  • Instruction leaflet