Drug test kit
Cocaine reagent
Jump to:
Discontinued
Product Details & Documents
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
- Kit comprises:
- 20 (4 boxes of 5, 0,2 ml) ampoules of Marquis reagent
- 1×10 ml cocaine reagent
- 1×10 ml barbiturate reagent
- 1×10 ml LSD reagent
- 1×20 ml cannabis reagent 1
- 1×10 ml cannabis reagent 2
- 1×5 g cannabis special solid reagent
- 1×20 g neutraliser
- 4 boxes of Whatman filter papers
- 1 microspatula
- 20 ampoule breakers
- 25 disposable spatulas
- forceps
- 4 watch glasses
- Instruction leaflet
Documents
Specifications
- Description:Cocaine reagent
- Seq:05