1-Hexene 97%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:592-41-6
- Formula:C₆H₁₂
- MDL Number:MFCD00009505
- Molecular Weight:84.16 g/mol
- Boiling Pt:62…65 °C (760 mmHg)
- Melting Pt:–140 °C
- Density:0.678 g/cm³
- Flash Pt:–26 °C
- Storage Temperature:Ambient
- EINECS:209-753-1
- UN:2370
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3867 to 1.3887 (20°C, 589 nm)
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 mL



