Kojic acid 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone
- CAS Number:501-30-4
- Formula:C₆H₆O₄
- MDL Number:MFCD00006580
- Molecular Weight:142.11 g/mol
- Merck Index:13,05334
- Boiling Pt:323 °C (1013 hPa)
- Melting Pt:151…156 °C
- Density:1,58 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-922-4
Specifications
Specification Test Results
- Appearance:White to beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:151 to 156 °C
- Titration with NaOH:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



