Order Entry
Italy
Orders LinkContactUsLinkComponent
Manganese(IV) oxide 88%, electrolytically precipitated, active

Manganese(IV) oxide 88%, electrolytically precipitated, active

Glass bottle for solids, 100 g
Catalog # ACRO203191000
Oxidizing
Irritant
Health
Manganese(IV) oxide 88%, electrolytically precipitated, active
Catalog # ACRO203191000
CAS Number:  1313-13-9
Manganese(IV) oxide 88%, electrolytically precipitated, active
Manganese(IV) oxide 88%, electrolytically precipitated, active

Glass bottle for solids, 100 g

CAS Number:  1313-13-9
Price
30,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
  • CAS Number:1313-13-9
  • Formula:MnO₂
  • MDL Number:MFCD00003463
  • Molecular Weight:86.94 g/mol
  • Merck Index:14,05730
  • Melting Pt:535 °C (dec.)
  • Density:5,03 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-202-6
  • UN:1479
  • ADR:5.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Black Powder
  • Titration Iodometric:
    ≥83 % (on dried substance)
  • Additional info:
    Oxidation activity: >60% (yield)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications