(±)-1,2-Octanediol 98+%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
(±)-1,2-Octanediol 98+%
Catalog # ACRO200140100
Supplier: Thermo Fisher Scientific
CAS Number: 1117-86-8
Jump to:
Product Details & Documents
Synonyms: 1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
- CAS Number:1117-86-8
- Formula:C₈H₁₈O₂
- MDL Number:MFCD00010738
- Molecular Weight:146.23 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:28…31 °C
- Flash Pt:138 °C
- EINECS:214-254-7
Documents
Specifications
Specification Test Results
- Appearance:Colorless to white Low melting solid
- Infrared spectrum:Conforms
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



