Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Selenium dioxide 99.8% (trace metals basis)

Glass bottle for solids, 500 g
Catalog # ACRO200005000
Toxic
Health
Environment
Selenium dioxide 99.8% (trace metals basis)
Catalog # ACRO200005000
CAS Number:  7446-08-4
Selenium dioxide 99.8% (trace metals basis)
Selenium dioxide 99.8% (trace metals basis)

Glass bottle for solids, 500 g

CAS Number:  7446-08-4
Price
386,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:13,08508
  • Melting Pt:315 °C (subl.)
  • Density:3,95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II

 

Specifications

Specification Test Results

  • Appearance:
    White to light beige Powder
  • Assay:
    70 to 71.5 % Se
  • Total trace metal impurities:
    ≤0.2 %
  • Arsenic (As):
    ≤20 ppm
  • Cadmium (Cd):
    ≤5 ppm
  • Copper (Cu):
    ≤10 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm
  • Mercury (Hg):
    ≤5 ppm
  • Nickel (Ni):
    ≤10 ppm
  • Tellurium (Te):
    ≤500 ppm
  • Insoluble matter:
    ≤0.2 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications