Order Entry
Italy
Orders LinkContactUsLinkComponent
Selenium dioxide 99.8% (trace metals basis)

Selenium dioxide 99.8% (trace metals basis)

Glass bottle for solids, 100 g
Catalog # ACRO200001000
Toxic
Health
Environment
Selenium dioxide 99.8% (trace metals basis)
Catalog # ACRO200001000
CAS Number:  7446-08-4
Selenium dioxide 99.8% (trace metals basis)
Selenium dioxide 99.8% (trace metals basis)

Glass bottle for solids, 100 g

CAS Number:  7446-08-4
Price
116,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:13,08508
  • Melting Pt:315 °C (subl.)
  • Density:3,95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light beige Powder
  • Assay:
    70 to 71.5 % Se
  • Total trace metal impurities:
    ≤0.2 %
  • Arsenic (As):
    ≤20 ppm
  • Cadmium (Cd):
    ≤5 ppm
  • Copper (Cu):
    ≤10 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm
  • Mercury (Hg):
    ≤5 ppm
  • Nickel (Ni):
    ≤10 ppm
  • Tellurium (Te):
    ≤500 ppm
  • Insoluble matter:
    ≤0.2 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.