Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Silver nitrate 99.85% for analysis

Glass bottle for solids, 25 g
Catalog # ACRO197680250
Oxidizing
Corrosive
Environment
Silver nitrate 99.85% for analysis
Catalog # ACRO197680250
CAS Number:  7761-88-8
Silver nitrate 99.85% for analysis
Silver nitrate 99.85% for analysis

Glass bottle for solids, 25 g

CAS Number:  7761-88-8
Price
133,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (760 mmHg)
  • Melting Pt:212 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43

 

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Loss on drying:
    ≤0.1 % (1 g, 70°C, 2 h)
  • Titration with HCl:
    ≥99.80 %
  • Chloride (Cl):
    ≤5 ppm
  • Nitrite (NO2):
    ≤10 ppm
  • Cadmium (Cd):
    ≤1 ppm
  • Calcium (Ca):
    ≤10 ppm
  • Chromium (Cr):
    ≤1 ppm
  • Copper (Cu):
    ≤1 ppm
  • Iron (Fe):
    ≤1 ppm
  • Lead (Pb):
    ≤1 ppm
  • Manganese (Mn):
    ≤1 ppm
  • Nickel (Ni):
    ≤1 ppm
  • Zinc (Zn):
    ≤1 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications