Lithium perchlorate trihydrate 99%, extra pure
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Lithium perchlorate trihydrate 99%, extra pure
Catalog # ACRO194722500
Supplier: Thermo Fisher Scientific
CAS Number: 13453-78-6
Jump to:
Product Details & Documents
- CAS Number:13453-78-6
- Formula:ClLiO₄·3H₂O
- MDL Number:MFCD00149765
- Molecular Weight:160.44 g/mol
- Melting Pt:95 °C
- Density:1,841 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-237-2
- UN:1481
- ADR:5.1,II
- :Density: 1.841 g/cm³ (20 °C)
Documents
ACRO19472
SDS
Specifications
Specification Test Results
- Appearance:White Adhering crystals and/or chunks
- Titration Argentometric:≥98.5 %
- Chloride (Cl):≤30 ppm
- Sulfate (SO4):≤10 ppm
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pack type:Plastic bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



