Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Selenium dioxide 99.999% (trace metals basis)

Glass bottle for solids, 50 g
Catalog # ACRO193980500
Toxic
Health
Environment
Selenium dioxide 99.999% (trace metals basis)
Catalog # ACRO193980500
CAS Number:  7446-08-4
Selenium dioxide 99.999% (trace metals basis)
Selenium dioxide 99.999% (trace metals basis)

Glass bottle for solids, 50 g

CAS Number:  7446-08-4
Price
491,00 €
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:13,08508
  • Melting Pt:315 °C
  • Density:3,95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II

 

Specifications

Specification Test Results

  • Appearance:
    Off-white Crystalline powder
  • Total trace metal impurities:
    ≤0.001 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications