Dimethyl adipate 99+%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Dimethyl adipate 99+%
Catalog # ACRO181315000
Supplier: Thermo Fisher Scientific
CAS Number: 627-93-0
Jump to:
Product Details & Documents
Synonyms: Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester
- CAS Number:627-93-0
- Formula:C₈H₁₄O₄
- MDL Number:MFCD00008469
- Molecular Weight:174.2 g/mol
- Merck Index:13,00163
- Boiling Pt:109…110 °C (14 mmHg)
- Melting Pt:8 °C
- Density:1.06 g/cm³
- Flash Pt:107 °C
- Storage Temperature:Ambient
- EINECS:211-020-6
Documents
ACRO18131
Scheda di sicurezza
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4270 to 1.4290 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Color scale:≤20 APHA
- Acidity:≤0.01 % (as adipic acid)
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



