Order Entry
Italy
Orders LinkContactUsLinkComponent
DL-Proline 99%

DL-Proline 99%

Glass bottle for solids, 5 g
Catalog # ACRO174030050
DL-Proline 99%
Catalog # ACRO174030050
CAS Number:  609-36-9
DL-Proline 99%
DL-Proline 99%

Glass bottle for solids, 5 g

CAS Number:  609-36-9
Price
54,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (±)-Proline, H-DL-Pro-OH
  • CAS Number:609-36-9
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00005250
  • Molecular Weight:115.13 g/mol
  • Merck Index:12,07963
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:208…210 °C
  • Density:1,38 g/cm³ (22.9 °C)
  • EINECS:210-189-3

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.5 % (K.F.)
  • Sulfated ash:
    ≤0.1 %
  • Solubility:
    (5 % in water) Colorless to light yellow
  • Titration with HClO4:
    98.5 to 101.5 %
  • Heavy metals (as Pb):
    ≤20 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications