Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Nitromethane 96% for spectroscopy

Glass bottle, 500 mL
Catalog # ACRO167865000
Flammable
Irritant
Health
Nitromethane 96% for spectroscopy
Catalog # ACRO167865000
CAS Number:  75-52-5
Nitromethane 96% for spectroscopy
Nitromethane 96% for spectroscopy

Glass bottle, 500 mL

CAS Number:  75-52-5
Price
101,00 €
Log in for availability
1 * 500 mL
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:101.2 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II

 

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 %
  • GC:
    ≥95 %
  • UV:
    at 380 nm A: ≤1
  • Color scale:
    ≤10 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.