Order Entry
Italy
Orders LinkContactUsLinkComponent
 

β-Alanine 99%

Plastic bottle, 500 g
Catalog # ACRO163795000
β-Alanine 99%
Catalog # ACRO163795000
CAS Number:  107-95-9
β-Alanine 99%
β-Alanine 99%

Plastic bottle, 500 g

CAS Number:  107-95-9
Price
72,20 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00204
  • Boiling Pt:201 °C (1013 hPa)
  • Melting Pt:197…202 °C (dec.)
  • Density:1,44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-536-5

 

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (105°C, 3 h)
  • Sulfated ash:
    ≤0.2 %
  • pH:
    6.5 to 7.5 (10 % w/v in water)
  • Titration with HClO4:
    ≥98.5 % (on dry substance)
  • Chloride (Cl):
    ≤400 ppm
  • Sulfate (SO4):
    ≤480 ppm
  • Ammonium (NH4):
    ≤1000 ppm
  • Iron (Fe):
    ≤30 ppm
  • Arsenic oxide (As2O3):
    ≤2 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    (TLC) Passes test (not detectable on 10 µg spotting)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.