Order Entry
Italy
Orders LinkContactUsLinkComponent
β-Alanine 99%

β-Alanine 99%

Plastic bottle, 2,5 kg
Catalog # ACRO163790025
β-Alanine 99%
Catalog # ACRO163790025
CAS Number:  107-95-9
β-Alanine 99%
β-Alanine 99%

Plastic bottle, 2,5 kg

CAS Number:  107-95-9
Price
258,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00204
  • Boiling Pt:201 °C (1013 hPa)
  • Melting Pt:197…202 °C (dec.)
  • Density:1,44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-536-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (105°C, 3 h)
  • Sulfated ash:
    ≤0.2 %
  • pH:
    6.5 to 7.5 (10 % w/v in water)
  • Titration with HClO4:
    ≥98.5 % (on dry substance)
  • Chloride (Cl):
    ≤400 ppm
  • Sulfate (SO4):
    ≤480 ppm
  • Ammonium (NH4):
    ≤1000 ppm
  • Iron (Fe):
    ≤30 ppm
  • Arsenic oxide (As2O3):
    ≤2 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    (TLC) Passes test (not detectable on 10 µg spotting)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications