Order Entry
Italy
Orders LinkContactUsLinkComponent
Pipette tips, universal, Corning®
Pipette tips, universal, Corning®
Beveled orifice, round, 1 - 200 µl, +, Transparent, 10 stack racks, 480 each
Catalog # 613-4240
   Low Manufacturing Impact
Supplier:   Corning
Pipette tips, universal, Corning®
Catalog # 613-4240
Supplier:  Corning
   Low Manufacturing Impact
Pipette tips, universal, Corning®
Pipette tips, universal, Corning®
Beveled orifice, round, 1 - 200 µl, +, Transparent, 10 stack racks, 480 each
Price
359,00 €
Log in for availability
1 * 4.800 pezzi
Product Details & Documents
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Specifications
  • Packed:
    10 stack racks, 480 each
  • Sterile:
    +
  • Volume:
    1 - 200 µl
  • Seq:
    02
  • Tip style:
    Beveled orifice, round
  • Tip colour:
    Transparent
  • Cat. No.:
    613-4240
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact