Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Octadecanol 95%

1-Octadecanol 95%

Plastic bottle, 1 kg
Catalog # ACRO129300010
1-Octadecanol 95%
Catalog # ACRO129300010
CAS Number:  112-92-5
1-Octadecanol 95%
1-Octadecanol 95%

Plastic bottle, 1 kg

CAS Number:  112-92-5
Price
50,40 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
  • CAS Number:112-92-5
  • Formula:H₃C(CH₂)₁₆CH₂OH
  • MDL Number:MFCD00002823
  • Molecular Weight:270.5 g/mol
  • Merck Index:12,08960
  • Boiling Pt:210 °C (15 mmHg)
  • Melting Pt:55…58 °C
  • Density:0,805 to 0.815 g/cm³ (60 °C)
  • Flash Pt:185 °C
  • Storage Temperature:Ambient
  • EINECS:204-017-6

Documents

Specifications

Specification Test Results

  • Appearance:
    White Powder or flakes or pellets
  • Infrared spectrum:
    Conforms
  • Melting point:
    55 to 60 °C
  • GC:
    ≥94 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications