Order Entry
Italy
Orders LinkContactUsLinkComponent
Nicotinamide 99%

Nicotinamide 99%

Plastic bottle, 100 g
Catalog # ACRO128271000
Irritant
Nicotinamide 99%
Catalog # ACRO128271000
CAS Number:  98-92-0
Nicotinamide 99%
Nicotinamide 99%

Plastic bottle, 100 g

CAS Number:  98-92-0
Price
38,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006395
  • Molecular Weight:122.13 g/mol
  • Merck Index:12,06574
  • Boiling Pt:150…160 °C (760 mmHg)
  • Melting Pt:128…131 °C
  • Density:1,4463 g/cm³ (20 °C)
  • Flash Pt:>150 °C
  • Storage Temperature:Ambient
  • EINECS:202-713-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    128 to 131 °C
  • Loss on drying:
    ≤0.5 % (4 hrs) (silica gel)
  • Sulfated ash:
    ≤0.1 %
  • Readily carbonizable substance:
    Passes test
  • pH:
    6.0 to 7.5 (5 % at 20 °C)
  • Titration with HClO4:
    ≥98.5 % (on dried substance)
  • Heavy metals (as Pb):
    ≤30 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications