Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
 

3-Mercaptopropionic acid 99+%

Bottiglia di vetro, 500 mL
  ACRO125535000
 
 
3-Mercaptopropionic acid 99+%
  ACRO125535000
 :  107-96-0
3-Mercaptopropionic acid 99+%
3-Mercaptopropionic acid 99+%

Bottiglia di vetro, 500 mL

 :  107-96-0
 
153,00 €
   
1 * 500 mL
 
  •  :107-96-0
  •  :C₃H₆O₂S
  •  :MFCD00004897
  •  :106.15 g/mol
  •  :110…111 °C (15 mmHg)
  •  :17…19 °C
  •  :1,218 g/cm³
  •  :124 °C
  •  :Ambiente
  •  :203-537-0
  •  :2922
  •  :8,II

 

 

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4910 to 1.4940 (20°C, 589 nm)
  • Titration Iodimetric:
    99.0 to 101.0 %

 

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    500 mL