3-Mercaptopropionic acid 99+%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
3-Mercaptopropionic acid 99+%
Catalog # ACRO125531000
Supplier: Thermo Fisher Scientific
CAS Number: 107-96-0
Jump to:
Product Details & Documents
- CAS Number:107-96-0
- Formula:C₃H₆O₂S
- MDL Number:MFCD00004897
- Molecular Weight:106.15 g/mol
- Boiling Pt:110…111 °C (15 mmHg)
- Melting Pt:17…19 °C
- Density:1.218 g/cm³
- Flash Pt:124 °C
- Storage Temperature:Ambient
- EINECS:203-537-0
- UN:2922
- ADR:8,II
Documents
ACRO12553
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4910 to 1.4940 (20°C, 589 nm)
- Titration Iodimetric:99.0 to 101.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



