Malonic acid 99%
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,3-Propanedioic acid
- CAS Number:141-82-2
- Formula:C₃H₄O₄
- MDL Number:MFCD00002707
- Molecular Weight:104.06 g/mol
- Merck Index:13,05732
- Boiling Pt:387 °C (1013 hPa)
- Melting Pt:130…135 °C (dec.)
- Density:1,669 g/cm³ (20 °C)
- Flash Pt:172 °C
- Storage Temperature:Ambient
- EINECS:205-503-0
Documents
ACRO12526
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:133 to 136 °C
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤500 ppm
- Sulfate (SO4):≤500 ppm
Specifications
- Pack type:Plastic bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



