1,6-Hexanedithiol 97%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
1,6-Hexanedithiol 97%
Catalog # ACRO120670250
Supplier: Thermo Fisher Scientific
CAS Number: 1191-43-1
Jump to:
Product Details & Documents
Synonyms: 1,6-Dimercaptohexane
- CAS Number:1191-43-1
- Formula:C₆H₁₄S₂
- MDL Number:MFCD00004910
- Molecular Weight:150.31 g/mol
- Boiling Pt:237 °C (760 mmHg)
- Melting Pt:–21 °C
- Density:0.99 g/cm³
- Flash Pt:90 °C
- Storage Temperature:Ambient
- EINECS:214-735-1
- UN:3334
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5110 to 1.5150 (20°C, 589 nm)
- GC:≥96.0 %
- Specific optical rotation:+16° to +18° (20°C, 589 nm) (c=1,H2O)
Specifications
- Pack type:Glass bottle
- Pk:25 g



