Order Entry
Italy
Orders LinkContactUsLinkComponent
1,6-Hexanedithiol 97%

1,6-Hexanedithiol 97%

Glass bottle for solids, 5 g
Catalog # ACRO120670050
1,6-Hexanedithiol 97%
Catalog # ACRO120670050
CAS Number:  1191-43-1
1,6-Hexanedithiol 97%
1,6-Hexanedithiol 97%

Glass bottle for solids, 5 g

CAS Number:  1191-43-1
Price
37,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1,6-Dimercaptohexane
  • CAS Number:1191-43-1
  • Formula:C₆H₁₄S₂
  • MDL Number:MFCD00004910
  • Molecular Weight:150.31 g/mol
  • Boiling Pt:237 °C (760 mmHg)
  • Melting Pt:–21 °C
  • Density:0.99 g/cm³
  • Flash Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:214-735-1
  • UN:3334
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5110 to 1.5150 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    +16° to +18° (20°C, 589 nm) (c=1,H2O)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications