1,6-Hexanediol 97%
Glass bottle for solids, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Hexamethylene glycol, 1,6-Dihydroxy hexane
- CAS Number:629-11-8
- Formula:C₆H₁₄O₂
- MDL Number:MFCD00002985
- Molecular Weight:118.18 g/mol
- Merck Index:13,04709
- Boiling Pt:253…260 °C (760 mmHg)
- Melting Pt:40…43 °C
- Density:0.96 g/cm³
- Flash Pt:147 °C
- Storage Temperature:Ambient
- EINECS:211-074-0
Specifications
Specification Test Results
- Appearance:Colorless to white to light yellow Solidified melt or liquid
- Infrared spectrum:Conforms
- Melting point:40 to 44 °C
- Water:≤0.3 % (K.F.)
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



