1,7-Heptanediol 95%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,7-Dihydroxy heptane
- CAS Number:629-30-1
- Formula:C₇H₁₆O₂
- MDL Number:MFCD00002987
- Molecular Weight:132.2 g/mol
- Boiling Pt:259 °C (760 mmHg)
- Melting Pt:17…19 °C
- Density:0.951 g/cm³
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:211-085-0
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4535 to 1.4565 (20°C, 589 nm)
- GC:≥94 %
- Specific optical rotation:-5.5° to -7.9° (20°C, 589 nm) (c=1,CHCl3)
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



