Sanger's reagent 98%
Glass bottle for solids, 25 g
: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene
- :70-34-8
- :C₆H₃FN₂O₄
- :MFCD00007056
- :186.1 g/mol
- :13,04199
- :296 °C
- :23…26 °C
- :1.48 g/cm³
- :>112 °C
- :Ambient
- :200-734-3
- :2811
- :6.1,II
- Appearance:Yellow to brown Liquid or low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.5675 to 1.5705 (20°C, 589 nm)
- GC:≥97.5 %
- Residue after ignition:≤0.05 %
- Suitability:(for precip. of Fe, etc.) Passes test
- Pack type:Glass bottle for solids
- Pk:25 g



